Learn More
Tris(4-methoxyphenyl)phosphine, 98%
CAS: 855-38-9 | C21H21O3P | 352.37 g/mol
$1112.14 - $1112.14
Chemical Identifiers
| CAS | 855-38-9 |
|---|---|
| Molecular Formula | C21H21O3P |
| Molecular Weight (g/mol) | 352.37 |
| MDL Number | MFCD00014896 |
| InChI Key | UYUUAUOYLFIRJG-UHFFFAOYSA-N |
| Synonym | tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine |
| PubChem CID | 70071 |
| IUPAC Name | tris(4-methoxyphenyl)phosphane |
| SMILES | COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1559614
|
Thermo Scientific Chemicals
A1559614 |
25 g |
Each for $1,112.14
|
|
|||||
Description
It is employed in Suzuki reaction. It is also used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed in Suzuki reaction. It is also used in the kinetic study of the effect of catalysts on the curing of biphenyl epoxy resin.
Solubility
Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 855-38-9 | |
| 352.37 | |
| UYUUAUOYLFIRJG-UHFFFAOYSA-N | |
| 70071 | |
| COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC |
| C21H21O3P | |
| MFCD00014896 | |
| tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine | |
| tris(4-methoxyphenyl)phosphane |
Specifications
| 855-38-9 | |
| C21H21O3P | |
| 25 g | |
| Air Sensitive | |
| Insoluble in water. | |
| COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC | |
| 352.37 | |
| 352.37 | |
| Tris(4-methoxyphenyl)phosphine |
| 130°C to 132°C | |
| MFCD00014896 | |
| 2815911 | |
| tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine | |
| UYUUAUOYLFIRJG-UHFFFAOYSA-N | |
| tris(4-methoxyphenyl)phosphane | |
| 70071 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 212-723-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only