missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-methoxyphenyl)phosphine, 95%, Thermo Scientific™

$76.92 - $578.23

Chemical Identifiers

CAS 855-38-9
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.36
MDL Number MFCD00014896
InChI Key UYUUAUOYLFIRJG-UHFFFAOYSA-N
Synonym tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine
PubChem CID 70071
IUPAC Name tris(4-methoxyphenyl)phosphane
SMILES COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC422240010
View Documents Promotion Details
N/A 1g Glass bottle
Each for $76.92
Only null left
 
AC422240100
View Documents Promotion Details
N/A 10g Glass bottle
Each for $578.23
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

855-38-9
352.36
UYUUAUOYLFIRJG-UHFFFAOYSA-N
70071
COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC
C21H21O3P
MFCD00014896
tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine
tris(4-methoxyphenyl)phosphane

Specifications

855-38-9
94% min. (GC)
MFCD00014896
Solubility in water: practically insoluble. Other solubilities: soluble in methanol
COC1=CC=C(C=C1)P(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)OC
352.36
352.36
Tris(4-methoxyphenyl)phosphine, 95%
Authentic
C21H21O3P
tris 4-methoxyphenyl phosphine, trianisylphosphine, tris p-anisyl phosphine, phosphine, tris 4-methoxyphenyl, tri-p-anisylphosphine, tris p-methoxyphenyl phosphine, tris 4-methoxyphenyl phosphane, phosphine, tris p-methoxyphenyl, tri 4-methoxyphenyl phosphine, tri p-methoxyphenyl phosphine
UYUUAUOYLFIRJG-UHFFFAOYSA-N
tris(4-methoxyphenyl)phosphane
70071
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 212-723-0

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only