Learn More
Tris(4-fluorophenyl)phosphine, 98+%, Thermo Scientific™
Chemical Identifiers
| CAS | 18437-78-0 |
|---|---|
| Molecular Formula | C18H12F3P |
| Molecular Weight (g/mol) | 316.26 |
| MDL Number | MFCD00013553 |
| InChI Key | GEPJPYNDFSOARB-UHFFFAOYSA-N |
| Synonym | tris 4-fluorophenyl phosphine, phosphine, tris 4-fluorophenyl, tris p-fluorophenyl phosphine, tri 4-fluorophenyl phosphine, tris 4-fluorophenyl phosphane, phosphine, tris p-fluorophenyl, zlchem 700, pubchem6418, acmc-1c3oi |
| PubChem CID | 140387 |
| IUPAC Name | tris(4-fluorophenyl)phosphane |
| SMILES | FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC306340010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
|
AC306340050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 18437-78-0 | |
| 316.26 | |
| GEPJPYNDFSOARB-UHFFFAOYSA-N | |
| 140387 | |
| FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1 |
| C18H12F3P | |
| MFCD00013553 | |
| tris 4-fluorophenyl phosphine, phosphine, tris 4-fluorophenyl, tris p-fluorophenyl phosphine, tri 4-fluorophenyl phosphine, tris 4-fluorophenyl phosphane, phosphine, tris p-fluorophenyl, zlchem 700, pubchem6418, acmc-1c3oi | |
| tris(4-fluorophenyl)phosphane |
Specifications
| 18437-78-0 | |
| 98% min. (GC) | |
| (FC6H4)3P | |
| tris 4-fluorophenyl phosphine, phosphine, tris 4-fluorophenyl, tris p-fluorophenyl phosphine, tri 4-fluorophenyl phosphine, tris 4-fluorophenyl phosphane, phosphine, tris p-fluorophenyl, zlchem 700, pubchem6418, acmc-1c3oi | |
| FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1 | |
| 316.26 | |
| 316.27 | |
| Tris(4-fluorophenyl)phosphine |
| Authentic | |
| C18H12F3P | |
| MFCD00013553 | |
| GEPJPYNDFSOARB-UHFFFAOYSA-N | |
| tris(4-fluorophenyl)phosphane | |
| 140387 | |
| 98+% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only