Learn More
Tris(4-fluorophenyl)phosphine, 98%
CAS: 18437-78-0 | C18H12F3P | 316.26 g/mol
Supplier: Thermo Scientific Chemicals A1481306
Description
Tris(4-fluorophenyl)phosphine, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTris(4-fluorophenyl)phosphine, is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.Stable under recommended storage conditions.
Specifications
| Tris(4-fluorophenyl)phosphine | |
| 78°C to 82°C | |
| MFCD00013553 | |
| 919838 | |
| tris 4-fluorophenyl phosphine, phosphine, tris 4-fluorophenyl, tris p-fluorophenyl phosphine, tri 4-fluorophenyl phosphine, tris 4-fluorophenyl phosphane, phosphine, tris p-fluorophenyl, zlchem 700, pubchem6418, acmc-1c3oi | |
| GEPJPYNDFSOARB-UHFFFAOYSA-N | |
| tris(4-fluorophenyl)phosphane | |
| 140387 | |
| 98% |
| 18437-78-0 | |
| C18H12F3P | |
| 5 g | |
| Air Sensitive | |
| Insoluble in water. | |
| FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1 | |
| 316.26 | |
| 316.27 |
Chemical Identifiers
| 18437-78-0 | |
| 316.26 | |
| GEPJPYNDFSOARB-UHFFFAOYSA-N | |
| 140387 | |
| FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1 |
| C18H12F3P | |
| MFCD00013553 | |
| tris 4-fluorophenyl phosphine, phosphine, tris 4-fluorophenyl, tris p-fluorophenyl phosphine, tri 4-fluorophenyl phosphine, tris 4-fluorophenyl phosphane, phosphine, tris p-fluorophenyl, zlchem 700, pubchem6418, acmc-1c3oi | |
| tris(4-fluorophenyl)phosphane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only