missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-bromophenyl)amine 98.0+%, TCI America™

Supplier:  TCI America T136125G

 View more versions of this product

Catalog No. T136125G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tris(4-bromophenyl)amine
146°C
C18H12Br3N
25 g
ZRXVCYGHAUGABY-UHFFFAOYSA-N
4-bromo-N,N-bis(4-bromophenyl)aniline
258027
≥98.0% (GC)
4316-58-9
White-Yellow
MFCD00009665
tris 4-bromophenyl amine, tris p-bromophenyl amine, tris-4-bromophenyl amine, 4-bromo-n,n-bis 4-bromophenyl aniline, benzenamine, 4-bromo-n,n-bis 4-bromophenyl, 4,4',4-tribromotriphenylamine, triphenylamine, 4,4',4-tribromo, pubchem13773, tri 4-bromophenyl amine, acmc-1arf9
C1=CC(=CC=C1N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br)Br
482.013
482.01
Crystalline Powder

Chemical Identifiers

4316-58-9
482.013
ZRXVCYGHAUGABY-UHFFFAOYSA-N
258027
C1=CC(=CC=C1N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br)Br
C18H12Br3N
MFCD00009665
tris 4-bromophenyl amine, tris p-bromophenyl amine, tris-4-bromophenyl amine, 4-bromo-n,n-bis 4-bromophenyl aniline, benzenamine, 4-bromo-n,n-bis 4-bromophenyl, 4,4',4-tribromotriphenylamine, triphenylamine, 4,4',4-tribromo, pubchem13773, tri 4-bromophenyl amine, acmc-1arf9
4-bromo-N,N-bis(4-bromophenyl)aniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions