accessibility menu, dialog, popup

UserName

BrNBrBr

CAS RN 4316-58-9

IUPAC Name: 4-bromo-N,N-bis(4-bromophenyl)aniline
Synonyms: 4-bromo-N,N-bis(4-bromophenyl)aniline
Molecular Weight (g/mol): 482.01
Molecular Formula: C18H12Br3N
InChi Key: ZRXVCYGHAUGABY-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N(C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)