missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-bromophenyl)amine 98.0+%, TCI America™

Catalog No. p-7136734
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13615G 5 g
T136125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13615G Supplier TCI America Supplier No. T13615G
Only null left

Chemical Identifiers

CAS 4316-58-9
Molecular Formula C18H12Br3N
Molecular Weight (g/mol) 482.013
MDL Number MFCD00009665
InChI Key ZRXVCYGHAUGABY-UHFFFAOYSA-N
Synonym tris 4-bromophenyl amine, tris p-bromophenyl amine, tris-4-bromophenyl amine, 4-bromo-n,n-bis 4-bromophenyl aniline, benzenamine, 4-bromo-n,n-bis 4-bromophenyl, 4,4',4-tribromotriphenylamine, triphenylamine, 4,4',4-tribromo, pubchem13773, tri 4-bromophenyl amine, acmc-1arf9
PubChem CID 258027
IUPAC Name 4-bromo-N,N-bis(4-bromophenyl)aniline
SMILES C1=CC(=CC=C1N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br)Br

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 5 g
Formula Weight 482.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-bromophenyl)amine