missing translation for 'onlineSavingsMsg'
Learn More

Tris(3-methoxyphenyl)phosphine, 98%

CAS: 29949-84-6 | C21H21O3P | 352.37 g/mol

$133.11 - $519.00

Chemical Identifiers

CAS 29949-84-6
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.37
MDL Number MFCD00008386
InChI Key CCXTYQMZVYIQRP-UHFFFAOYSA-N
Synonym tris 3-methoxyphenyl phosphine, tris 3-methoxyphenyl phosphane, tris m-methoxyphenyl phosphine, phosphine, tris 3-methoxyphenyl, ksc207e8l, tri m-anisyl phosphine, tris m-anisyl phosphine, phosphine,tris 3-methoxyphenyl, tris-3-methoxy-phenyl-phosphane, phosphine,tris 3-methoxybenzyl
PubChem CID 141534
IUPAC Name tris(3-methoxyphenyl)phosphane
SMILES COC1=CC(=CC=C1)P(C1=CC(OC)=CC=C1)C1=CC=CC(OC)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1794903
View Documents
Thermo Scientific Chemicals
A1794903
1 g
Each for $133.11
Only null left
 
AAA1794906
View Documents
Thermo Scientific Chemicals
A1794906
5 g
Each for $519.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

29949-84-6
352.37
CCXTYQMZVYIQRP-UHFFFAOYSA-N
141534
COC1=CC(=CC=C1)P(C1=CC(OC)=CC=C1)C1=CC=CC(OC)=C1
C21H21O3P
MFCD00008386
tris 3-methoxyphenyl phosphine, tris 3-methoxyphenyl phosphane, tris m-methoxyphenyl phosphine, phosphine, tris 3-methoxyphenyl, ksc207e8l, tri m-anisyl phosphine, tris m-anisyl phosphine, phosphine,tris 3-methoxyphenyl, tris-3-methoxy-phenyl-phosphane, phosphine,tris 3-methoxybenzyl
tris(3-methoxyphenyl)phosphane

Specifications

29949-84-6
C21H21O3P
1 g
tris 3-methoxyphenyl phosphine, tris 3-methoxyphenyl phosphane, tris m-methoxyphenyl phosphine, phosphine, tris 3-methoxyphenyl, ksc207e8l, tri m-anisyl phosphine, tris m-anisyl phosphine, phosphine,tris 3-methoxyphenyl, tris-3-methoxy-phenyl-phosphane, phosphine,tris 3-methoxybenzyl
COC1=CC(=CC=C1)P(C1=CC(OC)=CC=C1)C1=CC=CC(OC)=C1
352.37
352.37
Tris(3-methoxyphenyl)phosphine
113°C to 115°C
MFCD00008386
Air Sensitive
CCXTYQMZVYIQRP-UHFFFAOYSA-N
tris(3-methoxyphenyl)phosphane
141534
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only