CAS RN 29949-84-6
IUPAC Name:
tris(3-methoxyphenyl)phosphane
Synonyms:
tris(3-methoxyphenyl)phosphane
Molecular Weight (g/mol):
352.37
Molecular Formula:
C21H21O3P
InChi Key:
CCXTYQMZVYIQRP-UHFFFAOYSA-N
SMILES:
COC1=CC(=CC=C1)P(C1=CC(OC)=CC=C1)C1=CC=CC(OC)=C1
1
–
1
of
1
results
1
Promotions Available
Tris(3-methoxyphenyl)phosphine, 98%
Tris(3-methoxyphenyl)phosphine, 98%
CAS: 29949-84-6 Molecular Formula: C21H21O3P Molecular Weight (g/mol): 352.37 MDL Number: MFCD00008386 InChI Key: CCXTYQMZVYIQRP-UHFFFAOYSA-N Synonym: tris 3-methoxyphenyl phosphine,tris 3-methoxyphenyl phosphane,tris m-methoxyphenyl phosphine,phosphine, tris 3-methoxyphenyl,ksc207e8l,tri m-anisyl phosphine,tris m-anisyl phosphine,phosphine,tris 3-methoxyphenyl,tris-3-methoxy-phenyl-phosphane,phosphine,tris 3-methoxybenzyl PubChem CID: 141534 IUPAC Name: tris(3-methoxyphenyl)phosphane SMILES: COC1=CC(=CC=C1)P(C1=CC(OC)=CC=C1)C1=CC=CC(OC)=C1