missing translation for 'onlineSavingsMsg'
Learn More

Tris(3-methoxyphenyl)phosphine, 98%

Catalog No. AAA1794906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1794906 5 g
AAA1794903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1794906 Supplier Thermo Scientific Chemicals Supplier No. A1794906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29949-84-6 | C21H21O3P | 352.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29949-84-6
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.37
MDL Number MFCD00008386
InChI Key CCXTYQMZVYIQRP-UHFFFAOYSA-N
Synonym tris 3-methoxyphenyl phosphine, tris 3-methoxyphenyl phosphane, tris m-methoxyphenyl phosphine, phosphine, tris 3-methoxyphenyl, ksc207e8l, tri m-anisyl phosphine, tris m-anisyl phosphine, phosphine,tris 3-methoxyphenyl, tris-3-methoxy-phenyl-phosphane, phosphine,tris 3-methoxybenzyl
PubChem CID 141534
IUPAC Name tris(3-methoxyphenyl)phosphane
SMILES COC1=CC(=CC=C1)P(C1=CC(OC)=CC=C1)C1=CC=CC(OC)=C1

Specifications

Melting Point 113°C to 115°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 352.37
Percent Purity 98%
Chemical Name or Material Tris(3-methoxyphenyl)phosphine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only