missing translation for 'onlineSavingsMsg'
Learn More

Tris[3,5-bis(trifluoromethyl)phenyl]phosphine, 94%

CAS: 175136-62-6 | C24H9F18P | 670.281 g/mol

Supplier:  Thermo Scientific Chemicals L0694103

 View more versions of this product

Catalog No. AAL0694103

Only null left
Explore available promotions

Description

Description

Tris[3,5-bis(trifluoromethyl)phenyl]phosphine is used as ligands which is useful as catalysts in various reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine is used as ligands which is useful as catalysts in various reactions.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
98°C to 102°C
C24H9F18P
1 g
tris 3,5-bis trifluoromethyl phenyl phosphine, tris 3,5-bis trifluoromethyl phenyl phosphane, acmc-20ajat, tri 3,5-di trifluoromethyl phenyl phosphine, tris 3,5-bis-trifluoromethylphenyl phosphine, phosphine,tris 3,5-bis trifluoromethyl phenyl, tris 3,5-bis trifluoromethyl phenyl phosphine gc , yellow-brown
ITJHLZVYLDBFOJ-UHFFFAOYSA-N
tris[3,5-bis(trifluoromethyl)phenyl]phosphane
2778032
94%
175136-62-6
128°C to 130°C (0.5 mmHg)
MFCD00042428
6556841
Insoluble in water.
C1=C(C=C(C=C1C(F)(F)F)P(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C(F)(F)F
670.281
670.28

Chemical Identifiers

175136-62-6
670.281
ITJHLZVYLDBFOJ-UHFFFAOYSA-N
2778032
C1=C(C=C(C=C1C(F)(F)F)P(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C(F)(F)F
C24H9F18P
MFCD00042428
tris 3,5-bis trifluoromethyl phenyl phosphine, tris 3,5-bis trifluoromethyl phenyl phosphane, acmc-20ajat, tri 3,5-di trifluoromethyl phenyl phosphine, tris 3,5-bis-trifluoromethylphenyl phosphine, phosphine,tris 3,5-bis trifluoromethyl phenyl, tris 3,5-bis trifluoromethyl phenyl phosphine gc , yellow-brown
tris[3,5-bis(trifluoromethyl)phenyl]phosphane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only