missing translation for 'onlineSavingsMsg'
Learn More

Tris[3,5-bis(trifluoromethyl)phenyl]phosphine, 94%

Catalog No. p-7058317
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL06941MD 250 mg
AAL0694103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL06941MD Supplier Thermo Scientific Chemicals Supplier No. L06941MD
Only null left

CAS: 175136-62-6 | C24H9F18P | 670.281 g/mol

Tris[3,5-bis(trifluoromethyl)phenyl]phosphine is used as ligands which is useful as catalysts in various reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine is used as ligands which is useful as catalysts in various reactions.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 175136-62-6
Molecular Formula C24H9F18P
Molecular Weight (g/mol) 670.281
MDL Number MFCD00042428
InChI Key ITJHLZVYLDBFOJ-UHFFFAOYSA-N
Synonym tris 3,5-bis trifluoromethyl phenyl phosphine, tris 3,5-bis trifluoromethyl phenyl phosphane, acmc-20ajat, tri 3,5-di trifluoromethyl phenyl phosphine, tris 3,5-bis-trifluoromethylphenyl phosphine, phosphine,tris 3,5-bis trifluoromethyl phenyl, tris 3,5-bis trifluoromethyl phenyl phosphine gc , yellow-brown
PubChem CID 2778032
IUPAC Name tris[3,5-bis(trifluoromethyl)phenyl]phosphane
SMILES C1=C(C=C(C=C1C(F)(F)F)P(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 98°C to 102°C
Boiling Point 128°C to 130°C (0.5 mmHg)
Quantity 250 mg
Beilstein 6556841
Solubility Information Insoluble in water.
Formula Weight 670.28
Percent Purity 94%
Chemical Name or Material Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only