missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,4-dimethoxyphenyl)methanol, TCI America™

Catalog No. H07421G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07421G Supplier TCI America Supplier No. H07421G
Only null left

Chemical Identifiers

CAS 76832-37-6
Molecular Formula C25H28O7
Molecular Weight (g/mol) 440.49
MDL Number MFCD00145468
InChI Key SBBUIJKOSQBPNN-UHFFFAOYSA-N
Synonym Hexamethoxy Red, 2,2′C,2′C′C,4,4′C,4′C′C-Hexamethoxytriphenylcarbinol
PubChem CID 44630337
IUPAC Name tris(2,4-dimethoxyphenyl)methanol
SMILES COC1=CC(OC)=C(C=C1)C(O)(C1=C(OC)C=C(OC)C=C1)C1=C(OC)C=C(OC)C=C1

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 1 g
Formula Weight 440.49
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,4-dimethoxyphenyl)methanol