accessibility menu, dialog, popup

UserName

H3COOCH3OCH3OH3COH3COHOCH3

CAS RN 76832-37-6

IUPAC Name: tris(2,4-dimethoxyphenyl)methanol
Synonyms: tris(2,4-dimethoxyphenyl)methanol
Molecular Weight (g/mol): 440.49
Molecular Formula: C25H28O7
InChi Key: SBBUIJKOSQBPNN-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C=C1)C(O)(C1=C(OC)C=C(OC)C=C1)C1=C(OC)C=C(OC)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results