missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2,4-di-tert-butylphenyl) Phosphite 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 187°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 646.94 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tris(2,4-di-tert-butylphenyl) Phosphite |
Safety and Handling
| EINECSNumber | (3)-3510 |
| TSCA | Yes |