missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TRIS, 1.0M buffer soln., pH 8.0

Catalog No. AAJ62726K2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62726K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62726K2 Supplier Thermo Scientific™ Supplier No. J62726K2

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

Used as a biological buffer substance in the pH range 7—9; pKa = 8.3 at 20°C; pKa = 7.82 at 37°C. Used as a biological buffer or as a component of buffer formulations such as TAE and TBE buffers. Tris has a pKa of 8.06 and is very useful in the biology and biochemistry lab because it has buffering capabilities in the range of the typical physiological pH of most living organisms (pH 7.0-9.0). Tris has been reported to interfere with the activity of a number of enzymes, it should therefore be used carefully when studying proteins. Used in chemical manufacturing (surface-active agents, vulcanization accelerators, and pharmaceuticals), as an emulsifying agent (cosmetics, mineral oil and paraffin wax emulsions, leather dressings, textile specialties, polishes, cleaning compounds, and soluble oils), and as an absorbent for acidic gases, biological buffer, acidimetric standard, and therapeutic alkalinizing agent. Used in buffer systems for the freezer storage of sperm, to regulate pH in the transport of live aquatic species, cell culture media, and anti-inflammatory drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Concentration 1.0 M
pH 8.0
Physical Form Liquid
Quantity 1 L
Odor Odorless
Solubility Information Not miscible or difficult to mix in water. Soluble in ethylene glycol (79.1mg/ml),methanol (26mg/ml),anhydrous ethanol (14.6mg/ml),95% ethanol (22mg/ml),DMF (14mg/ml),acetone (2mg/ml),ethyl acetate (0.5mg/ml),olive oil (0.4mg/ml),chloroform (0.05mg/ml),and alkaline solution.
Chemical Name or Material TRIS buffer soln.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only