Learn More
TrippNT Balance Draft Shields
TrippNT Balance Draft Shields, Size: Medium, Dimensions: 15 x 15 x 15 in.
Supplier: TrippNT TRP50046
Description
TrippNT Balance Draft Shields, Size: Medium, Dimensions: 15 x 15 x 15 in.
Protect delicate instruments (balances and scales). Have top shield to help protect from down drafts and a mouse hole for power cords.
Specifications
| 18 x 18 x 21 in. | |
| Large | |
| Balance Draft Shield |