missing translation for 'onlineSavingsMsg'
Learn More

TrippNT Balance Draft Shields

Catalog No. p-3491135
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
Dimensions (L x W x H):
13 x 13 x 15 in.
15 x 15 x 15 in.
18 x 18 x 21 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Dimensions (L x W x H)
50855988 Small 13 x 13 x 15 in.
50855986 Large 18 x 18 x 21 in.
50855987 Medium 15 x 15 x 15 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50855988 Supplier TrippNT Supplier No. TRP 50029
Only null left

TrippNT Balance Draft Shields, Size: Medium, Dimensions: 15 x 15 x 15 in.

Protect delicate instruments (balances and scales). Have top shield to help protect from down drafts and a mouse hole for power cords.

Specifications

Dimensions (L x W x H) 13 x 13 x 15 in.
Size Small
Type Balance Draft Shield