Learn More
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringe
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
Supplier: Thermo Scientific™ 365I2351
Description
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
- Borosilicate glass with metal flanges
- Needles and plungers are stainless steel
- Field-proven syringe design
Specifications
| Stainless Steel (Needle), Stainless Steel (Plunger) | |
| 23 G | |
| GC Autosampler Syringe | |
| 250 μL | |
| Side Hole | |
| Fixed | |
| Yes |
| 250 μL FN GT, 23 G, 57 mm | |
| 57 mm | |
| Ea. | |
| 57 mm | |
| TriPlus RSH Autosampler | |
| Sterile |