missing translation for 'onlineSavingsMsg'
Learn More

Triphenylpropylphosphonium Bromide 98.0+%, TCI America™

Catalog No. p-7135004
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P120025G 25 g
P1200500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P120025G Supplier TCI America Supplier No. P120025G
Only null left

Chemical Identifiers

CAS 6228-47-3
Molecular Formula C21H22BrP
Molecular Weight (g/mol) 385.29
MDL Number MFCD00011843
InChI Key XMQSELBBYSAURN-UHFFFAOYSA-M
Synonym propyltriphenylphosphonium bromide, triphenyl propyl phosphonium bromide, 1-propyl triphenylphosphonium bromide, n-propyltriphenylphosphonium bromide, triphenyl propyl phosphanium bromide, n-propyl triphenylphosphonium bromide, propyl triphenyl phosphonium chloride, acmc-1bff2, propyltriphenylphosphoniumbromide
PubChem CID 80374
IUPAC Name triphenyl(propyl)phosphanium bromide
SMILES [Br-].CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 239°C
Color White
Quantity 25 g
Formula Weight 385.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylpropylphosphonium Bromide