accessibility menu, dialog, popup

UserName

BrCH3P

CAS RN 6228-47-3

IUPAC Name: triphenyl(propyl)phosphanium bromide
Synonyms: triphenyl(propyl)phosphanium bromide
Molecular Weight (g/mol): 385.29
Molecular Formula: C21H22BrP
InChi Key: XMQSELBBYSAURN-UHFFFAOYSA-M
SMILES: [Br-].CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)