missing translation for 'onlineSavingsMsg'
Learn More

(Triphenylphosphoranylidene)acetonitrile 98.0+%, TCI America™

Catalog No. T19585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19585G 5 g
T195825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19585G Supplier TCI America Supplier No. T19585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16640-68-9
Molecular Formula C20H16NP
Molecular Weight (g/mol) 301.33
MDL Number MFCD00567633
InChI Key APISVOVOJVZIBA-UHFFFAOYSA-N
Synonym triphenylphosphoranylidene acetonitrile, 2-triphenylphosphoranylidene acetonitrile, cyanomethylene triphenylphosphorane, acetonitrile, triphenylphosphoranylidene, 2-triphenyl-$l^ 5-phosphanylidene acetonitrile, 2-triphenyl-??-phosphanylidene acetonitrile, acmc-1c6hd, ksc493a4b, cyanomethylenetriphenylphosphorane
PubChem CID 85524
IUPAC Name 2-(triphenyl-λ⁵-phosphanylidene)acetonitrile
SMILES N#CC=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 193°C
Quantity 5 g
UN Number 2811
Formula Weight 301.33
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (Triphenylphosphoranylidene)acetonitrile