accessibility menu, dialog, popup

UserName

CAS RN 16640-68-9

IUPAC Name: 2-(triphenyl-λ⁵-phosphanylidene)acetonitrile
Synonyms: 2-(triphenyl-λ⁵-phosphanylidene)acetonitrile
Molecular Weight (g/mol): 301.33
Molecular Formula: C20H16NP
InChi Key: APISVOVOJVZIBA-UHFFFAOYSA-N
SMILES: N#CC=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)