missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphinecopper(I) hydride hexamer, 96%

CAS: 33636-93-0 | C108H96Cu6P6 | 1961.07 g/mol

$164.86 - $164.86

Chemical Identifiers

CAS 33636-93-0
Molecular Formula C108H96Cu6P6
Molecular Weight (g/mol) 1961.07
MDL Number MFCD00221518
InChI Key IZZFKTUYPRVQID-UHFFFAOYSA-T
Synonym stryker's reagent, triphenylphosphine copper hydride hexamer, triphenylphosphine-copper i hydride hexamer, unii-8x3r32m7xu, hydrido triphenylphosphine copper i hexamer, stryker's reagent mi, triphenylphosphine cuprous hydride, triphenylphosphine copper i hydride hexamer, copper, hexa-mu3-hydrohexakis triphenylphosphine hexa-, octahedro, cuprous hydride triphenylphosphine hexamer
PubChem CID 11982471
IUPAC Name copper;triphenylphosphane
SMILES [H+].[H+].[H+].[H+].[H+].[H+].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)[Cu]123[Cu]45([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]11([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]22([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]34([P+](C3=CC=CC=C3)(C3=CC=CC=C3)C3=CC=CC=C3)[Cu]512[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC382510010
SDS View Documents
Thermo Scientific Chemicals
382510010
1 g
Each for $164.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

33636-93-0
1961.07
IZZFKTUYPRVQID-UHFFFAOYSA-T
11982471
[H+].[H+].[H+].[H+].[H+].[H+].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)[Cu]123[Cu]45([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]11([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]22([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]34([P+](C3=CC=CC=C3)(C3=CC=CC=C3)C3=CC=CC=C3)[Cu]512[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C108H96Cu6P6
MFCD00221518
stryker's reagent, triphenylphosphine copper hydride hexamer, triphenylphosphine-copper i hydride hexamer, unii-8x3r32m7xu, hydrido triphenylphosphine copper i hexamer, stryker's reagent mi, triphenylphosphine cuprous hydride, triphenylphosphine copper i hydride hexamer, copper, hexa-mu3-hydrohexakis triphenylphosphine hexa-, octahedro, cuprous hydride triphenylphosphine hexamer
copper;triphenylphosphane

Specifications

33636-93-0
Authentic
Glass Bottle
MFCD00221518
15, 8987
Solubility in water: reacts slowly.
[H+].[H+].[H+].[H+].[H+].[H+].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)[Cu]123[Cu]45([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]11([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]22([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]34([P+](C3=CC=CC=C3)(C3=CC=CC=C3)C3=CC=CC=C3)[Cu]512[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1961.07
1961.07
Crystals
Red
96%
C108H96Cu6P6
1 g
stryker's reagent, triphenylphosphine copper hydride hexamer, triphenylphosphine-copper i hydride hexamer, unii-8x3r32m7xu, hydrido triphenylphosphine copper i hexamer, stryker's reagent mi, triphenylphosphine cuprous hydride, triphenylphosphine copper i hydride hexamer, copper, hexa-mu3-hydrohexakis triphenylphosphine hexa-, octahedro, cuprous hydride triphenylphosphine hexamer
IZZFKTUYPRVQID-UHFFFAOYSA-T
copper;triphenylphosphane
11982471
96%
Triphenylphosphinecopper(I) hydride hexamer
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only