missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine, powder, 99%

Catalog No. AAL0250218
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0250218 50 g
AAL0250230 250 g
AAL025020B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0250218 Supplier Thermo Scientific Chemicals Supplier No. L0250218

CAS: 603-35-0 | C18H15P | 262.29 g/mol

Triphenylphosphine is used in the synthesis of organic compounds due to its nucleophilicity and its reducing character. It is involved in the synthesis of biaryl compounds, phosphonium salts and other phosphorus compounds. As a reducing agent, it is used to prepare aromatic amines from the corresponding aromatic N-oxides. The anionic phosphine is usually isolated as the trisodium salt, which reacts with rhodium to form a complex that finds use in industrial hydroformylation reactions. It is also used to prepare Wilkinson's catalyst, RhCl(PPh3)3 useful to catalyze the hydrogenation of alkenes and tetrakis(triphenylphosphine)palladium(0) that is widely used to catalyse C-C coupling reactions in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylphosphine is used in the synthesis of organic compounds due to its nucleophilicity and its reducing character. It is involved in the synthesis of biaryl compounds, phosphonium salts and other phosphorus compounds. As a reducing agent, it is used to prepare aromatic amines from the corresponding aromatic N-oxides. The anionic phosphine is usually isolated as the trisodium salt, which reacts with rhodium to form a complex that finds use in industrial hydroformylation reactions. It is also used to prepare Wilkinson′s catalyst, RhCl(PPh3)3 useful to catalyze the hydrogenation of alkenes and tetrakis(triphenylphosphine)palladium(0) that is widely used to catalyse C-C coupling reactions in organic synthesis.

Solubility
Soluble in ether, benzene, carbon tetrachloride, glacial acetic acid, acetone, chloroform and alcohol. Insoluble in water.

Notes
Incompatible with oxidizing agents and acids.

Chemical Identifiers

CAS 603-35-0
Molecular Formula C18H15P
Molecular Weight (g/mol) 262.29
MDL Number MFCD00003043 MFCD20489348
InChI Key RIOQSEWOXXDEQQ-UHFFFAOYSA-N
Synonym triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resin
PubChem CID 11776
IUPAC Name triphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 79°C to 81°C
Density 1.2
Boiling Point 360°C
Flash Point 181°C (358°F)
Odor Mild
Quantity 50 g
Beilstein 610776
Merck Index 14,9743
Solubility Information Soluble in ether,benzene,carbon tetrachloride,glacial acetic acid,acetone,chloroform and alcohol. Insoluble in water.
Formula Weight 262.29
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Triphenylphosphine
Show More Show Less
Hazard Category H302-H317-H373
Hazard Statement GHS H Statement
H373-H302-H317
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P272-P280g-P301+P312-P302+P352-P314-P330-P333+P313-P363-P501c
EINECSNumber 210-036-0
RTECSNumber SZ3500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only