missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphine, 98%, Spectrum™ Chemical

Catalog No. 18614569
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
50 lb.
220 lb.
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614569 5 kg Amber Glass Bottle
18614568 25 g Amber Glass Bottle
18614570 250 g Amber Glass Bottle
18614572 50 lb. Amber Glass Bottle
18614571 220 lb. Amber Glass Bottle
18614567 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18614569 Supplier Spectrum Chemical Mfg Cor Supplier No. T11225KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 603-35-0

Specifications

CAS Max % 1
Boiling Point 360°C
Packaging Amber Glass Bottle
Quantity 5 kg
Percent Purity 0.98
Grade Reagent
Physical Form Crystals or Flakes

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.