missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethylium Tetrakis(pentafluorophenyl)borate 98.0+%, TCI America™

Catalog No. T28631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28631G 1 g
T28635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28631G Supplier TCI America Supplier No. T28631G

Chemical Identifiers

CAS 136040-19-2
Molecular Formula C43H15BF20
Molecular Weight (g/mol) 922.37
MDL Number MFCD03426981
InChI Key TZOSNOQHGGONMD-UHFFFAOYSA-N
Synonym triphenylmethylium tetrakis perfluorophenyl borate, trityl tetrakis pentafluorophenyl borate, trityl tetrapentafluorophenyl borate, methylium, triphenyl-, tetrakis pentafluorophenyl borate 1-, triphenylcarbenium tetrakis pentafluorophenyl borate, methylium, triphenyl-, tetrakis 2,3,4,5,6-pentafluorophenyl borate 1-1:1, tetrakis 2,3,4,5,6-pentafluorophenyl boranuide; triphenylmethylium, methylium, triphenyl-, tetrakis pentafluorophenyl borate 1-1:1, tritylium tetrakis pentafluorophenyl borate, triphenylmethyliumtetrakis pentafluorophenyl borate
PubChem CID 9832824
IUPAC Name tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide; triphenylmethylium
SMILES C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1.FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 183°C
Quantity 1 g
Formula Weight 922.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylmethylium Tetrakis(pentafluorophenyl)borate