missing translation for 'onlineSavingsMsg'
Learn More

Triphenylarsine, 97%

CAS: 603-32-7 | C18H15As | 306.24 g/mol

Supplier:  Thermo Scientific Chemicals L0361614

 View more versions of this product

Catalog No. AAL0361614

Only null left
Explore available promotions

Description

Description

Triphenylarsine is used in certain metal-catalyzed coupling reactions. It is a precursor to tetraphenylarsonium chloride a popular precipitating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylarsine is used in certain metal-catalyzed coupling reactions. It is a precursor to tetraphenylarsonium chloride a popular precipitating agent.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Handle and store under inert gas. Incompatible materials are Strong oxidizing agents.
Specifications

Specifications

Triphenylarsine
58°C to 62°C
265°C (509°F)
(C6H5)3As
25 g
1842037
Soluble in methanol. Insoluble in water.
C1=CC=C(C=C1)[As](C2=CC=CC=C2)C3=CC=CC=C3
306.24
306.24
603-32-7
360°C
C18H15As
MFCD00002994
UN3465
triphenylarsine, arsine, triphenyl, triphenyl arsenic, triphenylarsene, arsinetriphenyl, unii-mn8ez3fl74, mn8ez3fl74, triphenylarsenic, triphenyl arsine, triphenyl-arsine
BPLUKJNHPBNVQL-UHFFFAOYSA-N
triphenylarsane
11773
97%

Chemical Identifiers

603-32-7
306.24
BPLUKJNHPBNVQL-UHFFFAOYSA-N
11773
C1=CC=C(C=C1)[As](C2=CC=CC=C2)C3=CC=CC=C3
C18H15As
MFCD00002994
triphenylarsine, arsine, triphenyl, triphenyl arsenic, triphenylarsene, arsinetriphenyl, unii-mn8ez3fl74, mn8ez3fl74, triphenylarsenic, triphenyl arsine, triphenyl-arsine
triphenylarsane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.

P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c

H301+H331

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.

EINECSNumber : 210-032-9

RTECSNumber : CH7942500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only