missing translation for 'onlineSavingsMsg'
Learn More

Triphenylarsine, 97%

Catalog No. p-7043948
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0361606 5 g
AAL0361614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL0361606 Supplier Thermo Scientific Chemicals Supplier No. L0361606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 603-32-7 | C18H15As | 306.24 g/mol

Triphenylarsine is used in certain metal-catalyzed coupling reactions. It is a precursor to tetraphenylarsonium chloride a popular precipitating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylarsine is used in certain metal-catalyzed coupling reactions. It is a precursor to tetraphenylarsonium chloride a popular precipitating agent.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Handle and store under inert gas. Incompatible materials are Strong oxidizing agents.

Chemical Identifiers

CAS 603-32-7
Molecular Formula C18H15As
Molecular Weight (g/mol) 306.24
MDL Number MFCD00002994
InChI Key BPLUKJNHPBNVQL-UHFFFAOYSA-N
Synonym triphenylarsine, arsine, triphenyl, triphenyl arsenic, triphenylarsene, arsinetriphenyl, unii-mn8ez3fl74, mn8ez3fl74, triphenylarsenic, triphenyl arsine, triphenyl-arsine
PubChem CID 11773
IUPAC Name triphenylarsane
SMILES C1=CC=C(C=C1)[As](C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 58°C to 62°C
Boiling Point 360°C
Flash Point 265°C (509°F)
Linear Formula (C6H5)3As
Quantity 5 g
UN Number UN3465
Beilstein 1842037
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 306.24
Percent Purity 97%
Chemical Name or Material Triphenylarsine
Show More Show Less
Hazard Category H301+H331
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.
EINECSNumber 210-032-9
RTECSNumber CH7942500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only