accessibility menu, dialog, popup

UserName

CAS RN 603-32-7

IUPAC Name: triphenylarsane
Synonyms: triphenylarsine
Molecular Weight (g/mol): 306.24
Molecular Formula: C18H15As
InChi Key: BPLUKJNHPBNVQL-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[As](C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)