missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylamine 98.0+%, TCI America™
Supplier: TCI America T0507250G
Specifications
| Triphenylamine | |
| 127°C | |
| C18H15N | |
| 250 g | |
| ODHXBMXNKOYIBV-UHFFFAOYSA-N | |
| N,N-diphenylaniline | |
| 11775 | |
| ≥98.0% (GC) |
| 603-34-9 | |
| White-Yellow | |
| MFCD00003020 | |
| triphenylamine, benzenamine, n,n-diphenyl, triphenyl amine, n,n-diphenylbenzenamine, amine, triphenyl, unii-njs65m2ds2, ccris 4887, n,n,n-triphenylamine, n,n-diphenylbenzeneamine, njs65m2ds2 | |
| C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 245.325 | |
| 245.33 | |
| Crystalline Powder |
Chemical Identifiers
| 603-34-9 | |
| 245.325 | |
| ODHXBMXNKOYIBV-UHFFFAOYSA-N | |
| 11775 | |
| C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3 |
| C18H15N | |
| MFCD00003020 | |
| triphenylamine, benzenamine, n,n-diphenyl, triphenyl amine, n,n-diphenylbenzenamine, amine, triphenyl, unii-njs65m2ds2, ccris 4887, n,n,n-triphenylamine, n,n-diphenylbenzeneamine, njs65m2ds2 | |
| N,N-diphenylaniline |
Safety and Handling
RTECSNumber : YK2680000
TSCA : Yes