accessibility menu, dialog, popup

UserName

N

CAS RN 603-34-9

IUPAC Name: N,N-diphenylaniline
Synonyms: triphenylamine
Molecular Weight (g/mol): 245.33
Molecular Formula: C18H15N
InChi Key: ODHXBMXNKOYIBV-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)