missing translation for 'onlineSavingsMsg'
Learn More

Triphenylamine 98.0+%, TCI America™

Catalog No. p-7136317
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T050725G 25 g
T0507250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T050725G Supplier TCI America Supplier No. T050725G
Only null left

Chemical Identifiers

CAS 603-34-9
Molecular Formula C18H15N
Molecular Weight (g/mol) 245.325
MDL Number MFCD00003020
InChI Key ODHXBMXNKOYIBV-UHFFFAOYSA-N
Synonym triphenylamine, benzenamine, n,n-diphenyl, triphenyl amine, n,n-diphenylbenzenamine, amine, triphenyl, unii-njs65m2ds2, ccris 4887, n,n,n-triphenylamine, n,n-diphenylbenzeneamine, njs65m2ds2
PubChem CID 11775
IUPAC Name N,N-diphenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 25 g
Formula Weight 245.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylamine
RTECSNumber YK2680000
TSCA Yes