missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl(tetradecyl)phosphonium Bromide 98.0+%, TCI America™

Catalog No. T150625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T150625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T150625G Supplier TCI America Supplier No. T150625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25791-20-2
Molecular Formula C32H44BrP
Molecular Weight (g/mol) 539.58
MDL Number MFCD00051857
InChI Key FUMBGFNGBMYHGH-UHFFFAOYSA-M
Synonym triphenyl tetradecyl phosphonium bromide, myristyltriphenylphosphonium bromide, 1-tetradecyl triphenylphosphonium bromide, triphenyl tetradecyl phosphanium bromide, tetradecyl triphenylphosphonium bromide, acmc-209gmk, tetradecyltriphenylphosphoniumbromide, triphenyl tetradecyl phosphoniumbromide, n-tetradecyltriphenylphosphonium bromide, phosphonium,triphenyltetradecyl-, bromide 1:1
PubChem CID 3084442
IUPAC Name triphenyl(tetradecyl)phosphanium bromide
SMILES [Br-].CCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 94°C
Color White
Quantity 25 g
Formula Weight 539.58
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenyl(tetradecyl)phosphonium Bromide