accessibility menu, dialog, popup

UserName

BrH3CP

CAS RN 25791-20-2

IUPAC Name: triphenyl(tetradecyl)phosphanium bromide
Synonyms: triphenyl(tetradecyl)phosphanium bromide
Molecular Weight (g/mol): 539.58
Molecular Formula: C32H44BrP
InChi Key: FUMBGFNGBMYHGH-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)