missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl Borate 96.0+%, TCI America™

Catalog No. p-7118497
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B052425G 25 g
B0524500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B052425G Supplier TCI America Supplier No. B052425G
Only null left

Chemical Identifiers

CAS 1095-03-0
Molecular Formula C18H15BO3
Molecular Weight (g/mol) 290.125
MDL Number MFCD00059011
InChI Key MDCWDBMBZLORER-UHFFFAOYSA-N
Synonym triphenoxyborane, phenyl borate, triphenoxyboron, boric acid, triphenyl ester, phenyl borate, pho 3 b, trifenylester kyseliny borite, boric acid h3bo3 , triphenyl ester, triphenyl ester boric acid, trifenylester kyseliny borite czech, boric acid triphenyl ester
PubChem CID 14182
IUPAC Name triphenyl borate
SMILES B(OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3

Specifications

Melting Point 101°C
Color White-Yellow
Boiling Point 158°C
Quantity 25 g
UN Number 2811
Formula Weight 290.13
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenyl Borate
RTECSNumber ED5775000
TSCA Yes
Recommended Storage Refrigerator