accessibility menu, dialog, popup

UserName

CAS RN 1095-03-0

IUPAC Name: triphenyl borate
Synonyms: boric acid, triphenyl ester
Molecular Weight (g/mol): 290.13
Molecular Formula: C18H15BO3
InChi Key: MDCWDBMBZLORER-UHFFFAOYSA-N
SMILES: O(B(OC1=CC=CC=C1)OC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)