missing translation for 'onlineSavingsMsg'
Learn More

Triphenyl borate, 97%

CAS: 1095-03-0 | C18H15BO3 | 290.125 g/mol

$51.19 - $194.18

Chemical Identifiers

CAS 1095-03-0
Molecular Formula C18H15BO3
Molecular Weight (g/mol) 290.125
MDL Number MFCD00059011
InChI Key MDCWDBMBZLORER-UHFFFAOYSA-N
Synonym triphenoxyborane, phenyl borate, triphenoxyboron, boric acid, triphenyl ester, phenyl borate, pho 3 b, trifenylester kyseliny borite, boric acid h3bo3 , triphenyl ester, triphenyl ester boric acid, trifenylester kyseliny borite czech, boric acid triphenyl ester
PubChem CID 14182
IUPAC Name triphenyl borate
SMILES B(OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5568606
View Documents
Thermo Scientific Chemicals
H5568606
5 g
Each for $51.19
Only null left
 
AAH5568614
View Documents
Thermo Scientific Chemicals
H5568614
25 g
Each for $194.18
Only null left
 
Description

Description

Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyl borate is a useful boronic ester with chemosterilant activity against Cochliomyia hominivorax. It is also used as a competitive inhibitor of urease of Klebsiella aerogenes used to study the mechanisms of interaction with the nickel active site.

Solubility
Soluble in chloroform, methanol.

Notes
Moisture, water Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic. Store under inert atmosphere.
Specifications

Chemical Identifiers

1095-03-0
290.125
MDCWDBMBZLORER-UHFFFAOYSA-N
14182
B(OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3
C18H15BO3
MFCD00059011
triphenoxyborane, phenyl borate, triphenoxyboron, boric acid, triphenyl ester, phenyl borate, pho 3 b, trifenylester kyseliny borite, boric acid h3bo3 , triphenyl ester, triphenyl ester boric acid, trifenylester kyseliny borite czech, boric acid triphenyl ester
triphenyl borate

Specifications

1095-03-0
C18H15BO3
MFCD00059011
UN2811
triphenoxyborane, phenyl borate, triphenoxyboron, boric acid, triphenyl ester, phenyl borate, pho 3 b, trifenylester kyseliny borite, boric acid h3bo3 , triphenyl ester, triphenyl ester boric acid, trifenylester kyseliny borite czech, boric acid triphenyl ester
MDCWDBMBZLORER-UHFFFAOYSA-N
triphenyl borate
14182
97%
Triphenyl borate
98°C to 101°C
(C6H5O)3B
5 g
Moisture sensitive
Soluble in chloroform,methanol.
B(OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=CC=C3
290.125
290.13
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c

H301+H311-H315-H318-H330-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 214-137-0

RTECSNumber : ED5775000

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only