missing translation for 'onlineSavingsMsg'
Learn More

Trinitrophenol TS (Picric Acid TS), 1% (w/v), Ricca Chemical

Catalog No. 8696816
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8696816 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 8696816 Supplier Ricca Chemical Company Supplier No. 8696.816

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7732-18-5 , 88-89-1
Molecular Formula C6H3N3O7
Molecular Weight (g/mol) Mixture
InChI Key OXNIZHLAWKMVMX-UHFFFAOYSA-N
Synonym picric acid, trinitrophenol, carbazotic acid, picronitric acid, melinite, acide picrique, phenol trinitrate, picral, nitroxanthic acid, pikrinsaeure
PubChem CID 6954
ChEBI CHEBI:46149
IUPAC Name 2,4,6-trinitrophenol
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])[N+](=O)[O-]

Specifications

CAS Min % 97.0, 1.26
Color Yellow
CAS Max % 100.0, 1.31
Quantity 500 mL
Packaging Natural Poly Bottle
Physical Form Liquid
Percent Purity 1% w/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1% (w/v) Aqueous Solution
Chemical Name or Material Trinitrophenol TS (Picric Acid TS)
ShelfLife 24 months