missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsulfonium Hydroxide (0.2mol/L in Methanol) [Methylating Reagent], TCI America™
Supplier: TCI America T157625ML
Specifications
| Trimethylsulfonium Hydroxide (0.2mol/L in Methanol) [Methylating Reagent] | |
| Colorless | |
| MFCD00011632 | |
| 2924 | |
| VFJYIHQDILEQNR-UHFFFAOYSA-M | |
| trimethylsulfanium iodide | |
| 11105313 | |
| Liquid |
| 17287-03-5 | |
| C3H9IS | |
| 25 mL | |
| trimethylsulfonium hydroxide, trimethylsulfonium hydroxide solution, trimethylsulfonium hydroxide 0.2mol/l in methanol methylating reagent, sulfonium, trimethyl-, hydroxide, acmc-209sky, trimethylsulfoniumhydroxyd, ksc498e2f, trimethylsulfoniumhydroxidesolution, trimethylsulfonium hydroxide, 0.2mol/l methanol solution, trimethylsulfoniumhydroxide 0.2mol/linmethanol methylatingreagent | |
| [I-].C[S+](C)C | |
| 204.07 | |
| 94.17 |
Chemical Identifiers
| 17287-03-5 | |
| 204.07 | |
| VFJYIHQDILEQNR-UHFFFAOYSA-M | |
| 11105313 | |
| [I-].C[S+](C)C |
| C3H9IS | |
| MFCD00011632 | |
| trimethylsulfonium hydroxide, trimethylsulfonium hydroxide solution, trimethylsulfonium hydroxide 0.2mol/l in methanol methylating reagent, sulfonium, trimethyl-, hydroxide, acmc-209sky, trimethylsulfoniumhydroxyd, ksc498e2f, trimethylsulfoniumhydroxidesolution, trimethylsulfonium hydroxide, 0.2mol/l methanol solution, trimethylsulfoniumhydroxide 0.2mol/linmethanol methylatingreagent | |
| trimethylsulfanium iodide |
Safety and Handling
TSCA : No
Recommended Storage : Freezer