missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfonium Hydroxide (0.2mol/L in Methanol) [Methylating Reagent], TCI America™

Catalog No. p-7136845
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15765ML 5 mL
T157625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15765ML Supplier TCI America Supplier No. T15765ML
Only null left

Chemical Identifiers

CAS 17287-03-5
Molecular Formula C3H9IS
Molecular Weight (g/mol) 204.07
MDL Number MFCD00011632
InChI Key VFJYIHQDILEQNR-UHFFFAOYSA-M
Synonym trimethylsulfonium hydroxide, trimethylsulfonium hydroxide solution, trimethylsulfonium hydroxide 0.2mol/l in methanol methylating reagent, sulfonium, trimethyl-, hydroxide, acmc-209sky, trimethylsulfoniumhydroxyd, ksc498e2f, trimethylsulfoniumhydroxidesolution, trimethylsulfonium hydroxide, 0.2mol/l methanol solution, trimethylsulfoniumhydroxide 0.2mol/linmethanol methylatingreagent
PubChem CID 11105313
IUPAC Name trimethylsulfanium iodide
SMILES [I-].C[S+](C)C

Specifications

Color Colorless
Quantity 5 mL
UN Number 2924
Formula Weight 94.17
Physical Form Liquid
Chemical Name or Material Trimethylsulfonium Hydroxide (0.2mol/L in Methanol) [Methylating Reagent]
TSCA No
Recommended Storage Freezer