CAS RN 17287-03-5
CAS RN 17287-03-5
IUPAC Nor:
trimethylsulfanium iodide
Synonymes:
trimethylsulfonium iodide
Poids moléculaire (g/mol):
204.07
Formule moléculaire:
C3H9IS
InChi Key:
VFJYIHQDILEQNR-UHFFFAOYSA-M
SMILES:
[I-].C[S+](C)C
1
–
2
de
2
résultats
1
Trimethylsulfonium hydroxide, 0.25M solution in methanol
CAS: 17287-03-5 | C3H9IS | 204.07 g/mol
2
Trimethylsulfonium Hydroxide (0.2mol/L in Methanol) [Methylating Reagent], TCI America™
CAS: 17287-03-5 Formule moléculaire: C3H9IS Poids moléculaire (g/mol): 204.07 Numéro MDL: MFCD00011632 Clé InChI: VFJYIHQDILEQNR-UHFFFAOYSA-M Synonyme: trimethylsulfonium hydroxide,trimethylsulfonium hydroxide solution,trimethylsulfonium hydroxide 0.2mol/l in methanol methylating reagent,sulfonium, trimethyl-, hydroxide,acmc-209sky,trimethylsulfoniumhydroxyd,ksc498e2f,trimethylsulfoniumhydroxidesolution,trimethylsulfonium hydroxide, 0.2mol/l methanol solution,trimethylsulfoniumhydroxide 0.2mol/linmethanol methylatingreagent CID PubChem: 11105313 Nom IUPAC: trimethylsulfanium iodide SMILES: [I-].C[S+](C)C