missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylphenylammonium Bromide 98.0+%, TCI America™
Supplier: TCI America P0243500G
Specifications
| Trimethylphenylammonium Bromide | |
| White | |
| MFCD00011788 | |
| trimethylphenylammonium bromide, phenyltrimethylammonium bromide, n,n,n-trimethylbenzenaminium bromide, benzenaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylanilinium bromide, phenyl trimethyl ammonium bromide, ammonium, phenyltrimethyl-, bromide, trimethyl phenyl azanium bromide, benzenaminium, n,n,n-trimethyl-, bromide 1:1, ammonium trimethylphenyl-, bromide | |
| C[N+](C)(C)C1=CC=CC=C1.[Br-] | |
| 216.122 | |
| 216.12 | |
| Crystalline Powder |
| 16056-11-4 | |
| C9H14BrN | |
| 500 g | |
| GNMJFQWRASXXMS-UHFFFAOYSA-M | |
| trimethyl(phenyl)azanium;bromide | |
| 27663 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 16056-11-4 | |
| 216.122 | |
| GNMJFQWRASXXMS-UHFFFAOYSA-M | |
| 27663 | |
| C[N+](C)(C)C1=CC=CC=C1.[Br-] |
| C9H14BrN | |
| MFCD00011788 | |
| trimethylphenylammonium bromide, phenyltrimethylammonium bromide, n,n,n-trimethylbenzenaminium bromide, benzenaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylanilinium bromide, phenyl trimethyl ammonium bromide, ammonium, phenyltrimethyl-, bromide, trimethyl phenyl azanium bromide, benzenaminium, n,n,n-trimethyl-, bromide 1:1, ammonium trimethylphenyl-, bromide | |
| trimethyl(phenyl)azanium;bromide |
Safety and Handling
EINECSNumber : (9)-1187
RTECSNumber : BT2100000
TSCA : Yes