accessibility menu, dialog, popup

UserName

BrCH3NH3CCH3

CAS RN 16056-11-4

IUPAC Name: N,N,N-trimethylanilinium bromide
Synonyms: tetrabutylammonium tribromide bromide
Molecular Weight (g/mol): 216.12
Molecular Formula: C9H14BrN
InChi Key: GNMJFQWRASXXMS-UHFFFAOYSA-M
SMILES: [Br-].C[N+](C)(C)C1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)