Learn More
Trimethylboroxine, 50% w/w soln. in THF
CAS: 823-96-1 | C3H9B3O3 | 125.53 g/mol
Supplier: Thermo Scientific Chemicals H61464AC
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrimethylboroxine is used as a derivatizing agent for GLC analysis. It is used in a diverse array of areas, including as a polymerization additive. It is also used in the preparation of CBS catalysts for asymmetric reductions.
Solubility
Not miscible in water.
Notes
Moisture and light sensitive. Incompatible with oxidizing agents.
Specifications
| Trimethylboroxine | |
| 0.894 | |
| MFCD00013354 | |
| UN2924 | |
| Light and moisture sensitive | |
| Not miscible in water. | |
| CB1OB(C)OB(C)O1 | |
| 125.53 | |
| 125.54 |
| 823-96-1 | |
| C3H9B3O3 | |
| 25 mL | |
| 1757008 | |
| trimethylboroxine, 2,4,6-trimethylboroxin, trimethylboroxin, boroxin, trimethyl, trimethyl-1,3,5,2,4,6-trioxatriborinane, trimethyboroxine, trimethyl boroxin, trimethylboroxaine, trimethyl boroxine, trimethyl-boroxine | |
| GBBSAMQTQCPOBF-UHFFFAOYSA-N | |
| 2,4,6-trimethyl-1,3,5,2,4,6-trioxatriborinane | |
| 574072 | |
| 50% w/w soln. in THF |
Chemical Identifiers
| 823-96-1 | |
| 125.53 | |
| GBBSAMQTQCPOBF-UHFFFAOYSA-N | |
| 574072 | |
| CB1OB(C)OB(C)O1 |
| C3H9B3O3 | |
| MFCD00013354 | |
| trimethylboroxine, 2,4,6-trimethylboroxin, trimethylboroxin, boroxin, trimethyl, trimethyl-1,3,5,2,4,6-trioxatriborinane, trimethyboroxine, trimethyl boroxin, trimethylboroxaine, trimethyl boroxine, trimethyl-boroxine | |
| 2,4,6-trimethyl-1,3,5,2,4,6-trioxatriborinane |
Safety and Handling
GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P332+P313-P363-P370+P378q-P501c
H225-H315-H318-H335-H336-H351
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only