accessibility menu, dialog, popup

UserName

CAS RN 823-96-1

IUPAC Name: trimethyl-1,3,5,2,4,6-trioxatriborinane
Synonyms: trimethyl-1,3,5,2,4,6-trioxatriborinane
Molecular Weight (g/mol): 125.53
Molecular Formula: C3H9B3O3
InChi Key: GBBSAMQTQCPOBF-UHFFFAOYSA-N
SMILES: CB1OB(C)OB(C)O1
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)