missing translation for 'onlineSavingsMsg'
Learn More

Trimethylboroxine, 50% w/w soln. in THF

Catalog No. AAH61464AA
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61464AA 5 mL
AAH61464AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61464AA Supplier Thermo Scientific Chemicals Supplier No. H61464AA
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 823-96-1 | C3H9B3O3 | 125.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylboroxine is used as a derivatizing agent for GLC analysis. It is used in a diverse array of areas, including as a polymerization additive. It is also used in the preparation of CBS catalysts for asymmetric reductions.

Solubility
Not miscible in water.

Notes
Moisture and light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 823-96-1
Molecular Formula C3H9B3O3
Molecular Weight (g/mol) 125.53
MDL Number MFCD00013354
InChI Key GBBSAMQTQCPOBF-UHFFFAOYSA-N
Synonym trimethylboroxine, 2,4,6-trimethylboroxin, trimethylboroxin, boroxin, trimethyl, trimethyl-1,3,5,2,4,6-trioxatriborinane, trimethyboroxine, trimethyl boroxin, trimethylboroxaine, trimethyl boroxine, trimethyl-boroxine
PubChem CID 574072
IUPAC Name 2,4,6-trimethyl-1,3,5,2,4,6-trioxatriborinane
SMILES CB1OB(C)OB(C)O1

Specifications

Density 0.894
Quantity 5 mL
UN Number UN2924
Beilstein 1757008
Sensitivity Light and moisture sensitive
Solubility Information Not miscible in water.
Formula Weight 125.54
Concentration or Composition (by Analyte or Components) 50% w/w soln. in THF
Chemical Name or Material Trimethylboroxine
Hazard Category H225-H315-H318-H335-H336-H351
Hazard Statement GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only