missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl Orthobenzoate 95.0+%, TCI America™
Supplier: TCI America T228725G
Specifications
| Trimethyl Orthobenzoate | |
| Colorless | |
| C10H14O3 | |
| 25 g | |
| IECKAVQTURBPON-UHFFFAOYSA-N | |
| trimethoxymethylbenzene | |
| 69720 | |
| ≥95.0% (GC) |
| 707-07-3 | |
| 115°C | |
| MFCD00008474 | |
| trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate | |
| COC(C1=CC=CC=C1)(OC)OC | |
| 182.219 | |
| 182.22 | |
| Liquid |
Chemical Identifiers
| 707-07-3 | |
| 182.219 | |
| IECKAVQTURBPON-UHFFFAOYSA-N | |
| 69720 | |
| COC(C1=CC=CC=C1)(OC)OC |
| C10H14O3 | |
| MFCD00008474 | |
| trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate | |
| trimethoxymethylbenzene |
Safety and Handling
TSCA : Yes