missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl Orthobenzoate 95.0+%, TCI America™
$37.29 - $100.67
Chemical Identifiers
| CAS | 707-07-3 |
|---|---|
| Molecular Formula | C10H14O3 |
| Molecular Weight (g/mol) | 182.219 |
| MDL Number | MFCD00008474 |
| InChI Key | IECKAVQTURBPON-UHFFFAOYSA-N |
| Synonym | trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate |
| PubChem CID | 69720 |
| IUPAC Name | trimethoxymethylbenzene |
| SMILES | COC(C1=CC=CC=C1)(OC)OC |
Chemical Identifiers
| 707-07-3 | |
| 182.219 | |
| IECKAVQTURBPON-UHFFFAOYSA-N | |
| 69720 | |
| COC(C1=CC=CC=C1)(OC)OC |
| C10H14O3 | |
| MFCD00008474 | |
| trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate | |
| trimethoxymethylbenzene |
Specifications
| 707-07-3 | |
| 115°C | |
| MFCD00008474 | |
| trimethyl orthobenzoate, trimethoxymethyl benzene, methyl orthobenzoate, benzene, trimethoxymethyl, trimethylorthobenzoate, orthobenzoic acid, trimethyl ester, orthobenzoic acid trimethyl ester, alpha,alpha,alpha-trimethoxytoluene, trimethoxyphenylmethane, trimethyl orthobeznoate | |
| COC(C1=CC=CC=C1)(OC)OC | |
| 182.219 | |
| 182.22 | |
| Liquid |
| Colorless | |
| C10H14O3 | |
| 5 g | |
| IECKAVQTURBPON-UHFFFAOYSA-N | |
| trimethoxymethylbenzene | |
| 69720 | |
| ≥95.0% (GC) | |
| Trimethyl Orthobenzoate |
Safety and Handling
TSCA : Yes