missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethoxy[3-(phenylamino)propyl]silane 98.0+%, TCI America™
Supplier: TCI America P1458250G
Specifications
| Trimethoxy[3-(phenylamino)propyl]silane | |
| Yellow | |
| C12H21NO3Si | |
| 250 g | |
| n-3-trimethoxysilyl propyl aniline, trimethoxy 3-phenylamino propyl silane, n-phenylaminopropyltrimethoxysilane, unii-7au1h1fv6a, benzenamine, n-3-trimethoxysilyl propyl, n-phenyl-3-aminopropyltrimethoxysilane, 7au1h1fv6a, n-3-trimethoxysilylpropyl aniline, 3-phenylamino propyltrimethoxysilane | |
| CO[Si](CCCNC1=CC=CC=C1)(OC)OC | |
| 255.389 | |
| 255.39 | |
| Liquid |
| 3068-76-6 | |
| 310°C | |
| MFCD00053673 | |
| 3267 | |
| KBJFYLLAMSZSOG-UHFFFAOYSA-N | |
| N-(3-trimethoxysilylpropyl)aniline | |
| 76476 | |
| ≥98.0% (T) |
Chemical Identifiers
| 3068-76-6 | |
| 255.389 | |
| KBJFYLLAMSZSOG-UHFFFAOYSA-N | |
| 76476 | |
| CO[Si](CCCNC1=CC=CC=C1)(OC)OC |
| C12H21NO3Si | |
| MFCD00053673 | |
| n-3-trimethoxysilyl propyl aniline, trimethoxy 3-phenylamino propyl silane, n-phenylaminopropyltrimethoxysilane, unii-7au1h1fv6a, benzenamine, n-3-trimethoxysilyl propyl, n-phenyl-3-aminopropyltrimethoxysilane, 7au1h1fv6a, n-3-trimethoxysilylpropyl aniline, 3-phenylamino propyltrimethoxysilane | |
| N-(3-trimethoxysilylpropyl)aniline |
Safety and Handling
EINECSNumber : (3)-2644
TSCA : Yes